Butyl-p-toluenesulphonate ≥97.0%
Leverantör: TCI
T0727-25ML
T0727-500ML
TCIAT0727-25MLEA
495
SEK
TCIAT0727-25ML
TCIAT0727-500ML
Butyl-p-toluenesulphonate ≥97.0%
Butyl-p-toluenesulphonate
Formel:
C₁₁H₁₆O₃S Kokpunkt: 172°C |
MDL Number:
MFCD00027203 CAS nummer: 778-28-9 EINECS: 212-295-5 |
|
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...