2,4,6-Trimethylstyrene ≥97.0% stabilised
Leverantör: TCI
|
Varning
|
Stabilization:
med 4-tert-Butylpyrokatekol
T0493-1ML
T0493-5ML
TCIAT0493-1MLEA
470
SEK
TCIAT0493-1ML
TCIAT0493-5ML
2,4,6-Trimethylstyrene ≥97.0% stabilised
2,4,6-Trimethylstyrene
Formel:
C₁₁H₁₄ Kokpunkt: 206°C Flampunkt: 75°C Storage Temperature: Kylskåp |
MDL Number:
MFCD00008613 CAS nummer: 769-25-5 EINECS: 212-205-4 |
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...