m-Tolyldiethanolamine ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
2,2'-(3-Methylphenylimino)diethanol, 2,2'-(m-Tolylimino)diethanol, N,N-Bis(2-hydroxyethyl)-m-toluidine, 2,2'-(m-Tolylazanediyl)diethanol
T0315-25G
T0315-500G
TCIAT0315-25GEA
295
SEK
TCIAT0315-25G
TCIAT0315-500G
m-Tolyldiethanolamine ≥98.0%
m-Tolyldiethanolamine
Formel:
C₁₁H₁₇NO₂ MW: 195,26 g/mol |
MDL Number:
MFCD00020578 CAS nummer: 91-99-6 EINECS: 202-114-8 |
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 70.0 to 73.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...