6-Thioguanine ≥95.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 6-Mercaptoguanine, 2-Amino-6-purinethiol, 2-Amino-3,7-dihydro-6H-purine-6-thione, 2-Amino-6-mercaptopurine

T0212-1G T0212-5G
TCIAT0212-1GEA 645 SEK
TCIAT0212-1G TCIAT0212-5G
6-Thioguanine ≥95.0%
6-Thioguanine
Formel: C₅H₅N₅S
MW: 167,19 g/mol
MDL Number: MFCD00233553
CAS nummer: 154-42-7
EINECS: 205-827-2
UN: 2811
ADR: 6.1,III
Merck Index: 14,09337

Order Now

Specifikation testresultat

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 95.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR