5,5'-Thiodisalicylic acid ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
T0208-25G
T0208-5G
TCIAT0208-25GEA
3775
SEK
TCIAT0208-25G
TCIAT0208-5G
5,5'-Thiodisalicylic acid ≥98.0% (by GC, titration analysis)
5,5'-Thiodisalicylic acid
Formel:
C₁₄H₁₀O₆S |
MDL Number:
MFCD00020264 CAS nummer: 1820-99-1 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in hot EtOH | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...