1,2,3,4-Tetrahydroquinaldine 97,0 GC_ANALYSMETOD

Leverantör: TCI
Varning

Synonyms: 2-Methyl-1,2,3,4-tetrahydroquinoline, 1,2,3,4-Tetrahydro-2-methylquinoline

T0112-25G T0112-500G
TCIAT0112-25GEA 990 SEK
TCIAT0112-25G TCIAT0112-500G
1,2,3,4-Tetrahydroquinaldine 97,0 GC_ANALYSMETOD
1,2,3,4-Tetrahydroquinaldine
Formel: C₁₀H₁₃N
MW: 147.22 g/mol
Kokpunkt: 250°C
MDL Number: MFCD00023886
CAS nummer: 1780-19-4
EINECS: 217-226-2

Order Now

Specifikation testresultat

Appearance Colorless to Orange to Green clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR