Tetradecenylsuccinic anhydride ≥85.0% (by GC, titration analysis)

Leverantör: TCI
Varning

T0089-25G T0089-500G
TCIAT0089-25GEA 215 SEK
TCIAT0089-25G TCIAT0089-500G
Tetradecenylsuccinic anhydride ≥85.0% (by GC, titration analysis)
Tetradecenylsuccinic anhydride
Formel: C₁₈H₃₀O₃
MDL Number: MFCD00014543
CAS nummer: 33806-58-5
EINECS: 251-682-3

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 85.0 %
Purity (Morpholine Method) min. 85.0 %
Melting point 48.0 to 55.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR