Tetrachlorophthalic acid hemihydrate ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 3,4,5,6-Tetrachlorophthalic acid

T0070-25G T0070-500G
TCIAT0070-25GEA 470 SEK
TCIAT0070-25G TCIAT0070-500G
Tetrachlorophthalic acid hemihydrate ≥98.0%
Tetrachlorophthalic acid hemihydrate
Formel: C₈H₂Cl₄O₄
MW: 303,90 g/mol
MDL Number: MFCD00053309
CAS nummer: 632-58-6
EINECS: 211-181-2

Order Now

Specifikation testresultat

Appearance White powder to crystal
Purity (Neutralization titration) min. 98.0 %
Water 2.7 to 3.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR