Silver(I) hexafluorophosphate ≥98.0%

Leverantör: TCI
Fara

Synonyms: Silver hexafluorophosphate

S0981-1G S0981-5G
TCIAS0981-1GEA 405 SEK
TCIAS0981-1G TCIAS0981-5G
Silver(I) hexafluorophosphate ≥98.0%
Silver(I) hexafluorophosphate
Formel: AgF₆P
MDL Number: MFCD00003415
CAS nummer: 26042-63-7
EINECS: 247-428-6
UN: 3260
ADR: 8,II

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (Precipitation Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR