Seratrodast ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI
Varning
Synonyms: 7-Phenyl-7-(2,4,5-trimethyl-3,6-dioxocyclohexa-1,4-dien-1-yl)heptanoic acid
S0943-100MG S0943-1G
TCIAS0943-100MGEA 765 SEK
TCIAS0943-100MG TCIAS0943-1G
Seratrodast ≥98.0% (by HPLC, titration analysis)
Seratrodast
Formel: C₂₂H₂₆O₄
MW: 354.45 g/mol
Storage Temperature: Frys
MDL Number: MFCD00875701
CAS nummer: 112665-43-7
Merck Index: 14,08459
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52e5ca5d55d7b4ef9c2d806c743a316e01f6e00a&vid=52e5ca5d55d7b4ef9c2d806c743a316e01f6e00a&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 126.0 to 130.0 deg-C