(R)-(+)-SEGPHOS® ≥99.0%

Leverantör: TCI

Synonyms: (R)-(+)-4,4'-Bis(diphenylphosphino)-3,3'-bi(1,2-methylenedioxybenzene), (R)-(+)-SEGPHOS®, (R)-(+)-5,5'-Bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole

S0930-200MG S0930-1G
TCIAS0930-200MGEA 430 SEK
TCIAS0930-200MG TCIAS0930-1G
(R)-(+)-SEGPHOS® ≥99.0%
(R)-5,5'-Bis(diphenylphosphino)-4,4'-bibenzo[d][1,3]dioxole
Formel: C₃₈H₂₈O₄P₂
MW: 610.570 g/mol
MDL Number: MFCD09753005
CAS nummer: 244261-66-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9245aa46003cb573cc6bd40c08289da7255f5018&vid=9245aa46003cb573cc6bd40c08289da7255f5018&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 99.0 area%
Optical purity (LC) min. 99.0 ee%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR