Splitomicin ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 1,2-Dihydro-3H-naphtho[2,1-b]pyran-3-one, 1,2-Dihydro-3H-benzo[f]chromen-3-one

S0892-200MG S0892-1G
TCIAS0892-200MGEA 1050 SEK
TCIAS0892-200MG TCIAS0892-1G
Splitomicin ≥98.0%
Splitomicin
Formel: C₁₃H₁₀O₂
MW: 198,22 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD08705254
CAS nummer: 5690-03-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a67af9ab3a329ab697ab4e89dd6c1bfdba2ae607&vid=a67af9ab3a329ab697ab4e89dd6c1bfdba2ae607&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 53.0 to 57.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR