1-(3-sulphopropyl)pyridinium hydroxide inner salt ≥98.0% for biochemical research
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
NDSB 201, 3-(1-Pyridinio)propanesulphonate, 3-(Pyridin-1-ium-1-yl)propane-1-sulphonate
S0813-5G
S0813-25G
TCIAS0813-5GEA
285
SEK
TCIAS0813-5G
TCIAS0813-25G
1-(3-sulphopropyl)pyridinium hydroxide inner salt ≥98.0% for biochemical research
1-(3-sulphopropyl)pyridinium hydroxide inner salt
Formel:
C₈H₁₁NO₃S MW: 201,24 g/mol Smältpunkt: 275°C |
MDL Number:
MFCD00064468 CAS nummer: 15471-17-7 EINECS: 239-491-3 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Suitability for protein-analysis | to pass test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...