(S)-(-)-Epoxystyrene 98,0 GC_ANALYSMETOD
Leverantör: TCI
|
Fara
|
Synonyms:
(S)-Styrene oxide
S0517-25G
S0517-5G
TCIAS0517-25GEA
2150
SEK
TCIAS0517-25G
TCIAS0517-5G
(S)-(-)-Epoxystyrene 98,0 GC_ANALYSMETOD
(S)-(-)-Epoxystyrene
Formel:
C₈H₈O MW: 120.15 g/mol Kokpunkt: 89°C Flampunkt: 77°C |
MDL Number:
MFCD00064310 CAS nummer: 20780-54-5 UN: 2810 ADR: 6.1 III |
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...