(R)-(+)-Epoxystyrene 96,0 GC_ANALYSMETOD
Leverantör: TCI
|
Fara
|
Synonyms:
(R)-(+)-Styrene oxide, (R)-Styrene oxide
S0516-5G
S0516-25G
TCIAS0516-5GEA
815
SEK
TCIAS0516-5G
TCIAS0516-25G
(R)-(+)-Epoxystyrene 96,0 GC_ANALYSMETOD
(R)-(+)-Epoxystyrene
Formel:
C₈H₈O MW: 120.15 g/mol Kokpunkt: 194°C Flampunkt: 77°C |
MDL Number:
MFCD00066210 CAS nummer: 20780-53-4 UN: 2810 ADR: 6.1 III |
|
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 96.0 % |
Specific rotation [a]20/D | +31.0 to +34.0 deg (neat) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...