Succinaldehyde bis(dimethyl acetal) ≥98.0%

Leverantör: TCI

Synonyms: Succinaldehyde Tetramethyl Acetal, 1,1,4,4-Tetramethoxybutane

S0371-25G S0371-5G
TCIAS0371-25GEA 2150 SEK
TCIAS0371-25G TCIAS0371-5G
Succinaldehyde bis(dimethyl acetal) ≥98.0%
Succinaldehyde bis(dimethyl acetal)
Formel: C₈H₁₈O₄
Kokpunkt: 89°C
MDL Number: MFCD00127971
CAS nummer: 6922-39-0

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR