Trimethylstearylammonium chloride ≥98.0%

Leverantör: TCI
Fara

S0087-25G S0087-500G S0087-100G
TCIAS0087-25GEA 270 SEK
TCIAS0087-25G TCIAS0087-500G TCIAS0087-100G
Trimethylstearylammonium chloride ≥98.0%
Trimethylstearylammonium chloride
Formel: C₂₁H₄₆ClN
MDL Number: MFCD00050188
CAS nummer: 112-03-8
EINECS: 203-929-1
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=efbaffb734561056c43f2814c806da0edb40e4f6&vid=efbaffb734561056c43f2814c806da0edb40e4f6&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR