Stearamide ≥90.0%

Leverantör: TCI EUROPE

Synonyms: Octadecanamide, Stearic acid amide

S0075-25G S0075-250G
TCIAS0075-25GEA 215 SEK
TCIAS0075-25G TCIAS0075-250G
Stearamide ≥90.0%
Stearamide
Formel: C₁₈H₃₇NO
MW: 283,50 g/mol
MDL Number: MFCD00008038
CAS nummer: 124-26-5
EINECS: 204-693-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa2e2f20a5cdc02708202196991f1bf8b89b4b0d&vid=aa2e2f20a5cdc02708202196991f1bf8b89b4b0d&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 90.0 %
Melting point 104.0 to 111.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR