Spermidine phosphate ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
N-(3-Aminopropyl)-1,4-butanediamine Phosphate, N-(3-Aminopropyl)tetramethylenediamine Phosphate
S0068-1G
S0068-5G
TCIAS0068-1GEA
455
SEK
TCIAS0068-1G
TCIAS0068-5G
Spermidine phosphate ≥98.0%
Spermidine phosphate
Formel:
C₇H₁₉N₃ MW: 439,23 g/mol |
MDL Number:
MFCD00043283 CAS nummer: 49721-50-8 EINECS: 256-447-9 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...