RPI 1 ≥95.0%
Leverantör: TCI EUROPE
Synonyms:
1,3-Dihydro-5,6-dimethoxy-3-[(4-hydroxyphenyl)methylene]-1H-indol-2-one, 3-(4-Hydroxybenzylidene)-5,6-dimethoxyindolin-2-one, RPI-1
R0202-10MG
R0202-50MG
TCIAR0202-10MGEA
1125
SEK
TCIAR0202-10MG
TCIAR0202-50MG
RPI 1 ≥95.0%
RPI 1
Formel:
C₁₇H₁₅NO₄ MW: 297,31 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD03852474 CAS nummer: 269730-03-2 |
Specifikation testresultat
Appearance | Light yellow to Brown powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Elemental analysis (Nitrogen) | 4.20 to 5.20 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...