[RuCl(p-cymene)((S)-tolbinap)]Cl

Leverantör: TCI

Synonyms: Chloro[(S)-(-)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl](p-cymene)ruthenium(II) Chloride, Chloro[(S)-(-)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl](p-cymene)ruthenium(II) chloride [RuCl(p-cymene)((S)-tolbinap)]Cl

R0149-200MG R0149-1G
TCIAR0149-200MGEA 445 SEK
TCIAR0149-200MG TCIAR0149-1G
[RuCl(p-cymene)((S)-tolbinap)]Cl
[RuCl(p-cymene)((S)-tolbinap)]Cl
Formel: C₅₈H₅₄Cl₂P₂Ru
MDL Number: MFCD09753014
CAS nummer: 228120-95-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a75b5d915ce1b365cf25641660c866d796a8c68&vid=5a75b5d915ce1b365cf25641660c866d796a8c68&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Brown powder to crystal
IR to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR