[RuCl(p-cymene)((S)-binap)]Cl ≥90.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
(S)-RuCl[(p-cymene(BINAP)]Cl, Chloro[(S)-(-)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl](p-cymene)ruthenium(II) chloride [RuCl(p-cymene)((S)-binap)]Cl, Chloro[(S)-(-)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl](p-cymene)ruthenium(II) Chloride, [(S)-Binap RuCl p-cymene]Cl
R0147-200MG
R0147-1G
TCIAR0147-200MGEA
455
SEK
TCIAR0147-200MG
TCIAR0147-1G
[RuCl(p-cymene)((S)-binap)]Cl ≥90.0%
[RuCl(p-cymene)((S)-binap)]Cl
Formel:
C₅₄H₄₆Cl₄P₂Ru MW: 999,77 g/mol |
MDL Number:
MFCD00134456 CAS nummer: 130004-33-0 |
Specifikation testresultat
Appearance | White to Brown to Dark purple powder to crystal |
Purity (HPLC) | min. 90 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...