Regorafenib ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 4-(4-(3-(4-Chloro-3-(trifluoromethyl)phenyl)ureido)-3-fluorophenoxy)-N-methylpicolinamide, BAY-73-4506
R0142-25MG R0142-100MG
TCIAR0142-25MGEA 950 SEK
TCIAR0142-25MG TCIAR0142-100MG
Regorafenib ≥98.0%
Regorafenib
Formel: C₂₁H₁₅ClF₄N₄O₃
MW: 482,8 g/mol
Storage Temperature: Frys
MDL Number: MFCD16038047
CAS nummer: 755037-03-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e6020ef7c375ffd17eb9143aa76f8fcfff800c6&vid=3e6020ef7c375ffd17eb9143aa76f8fcfff800c6&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 11.00 to 12.20 %