RuCl2[(R)-xylbinap][(R)-daipen]

Leverantör: TCI EUROPE

Synonyms: Dichloro{(R)-(+)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl}[(2R)-(-)-1,1-bis(4-methoxyphenyl)-3-methyl-1,2-butanediamine]ruthenium(II) RuCl2[(R)-xylbinap][(R)-daipen]

R0133-200MG R0133-1G
TCIAR0133-200MGEA 590 SEK
TCIAR0133-200MG TCIAR0133-1G
RuCl2[(R)-xylbinap][(R)-daipen]
RuCl2[(R)-xylbinap][(R)-daipen]
Formel: C₇₁H₇₄Cl₂N₂O₂P₂Ru
MW: 1221,30 g/mol
MDL Number: MFCD09753026
CAS nummer: 220114-32-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31f795baa764f56a74c80169ea83b87790fdfbdd&vid=31f795baa764f56a74c80169ea83b87790fdfbdd&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Amber to Dark green powder to crystal
IR to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR