Ritalinic acid ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE

Synonyms: 2-Phenyl-2-(2-piperidyl)acetic acid

R0111-1G R0111-5G
TCIAR0111-1GEA 550 SEK
TCIAR0111-1G TCIAR0111-5G
Ritalinic acid ≥98.0% (by HPLC, titration analysis)
Ritalinic acid
Formel: C13H17NO2
MDL Number: MFCD06200695
CAS nummer: 19395-41-6
EINECS: 243-020-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33a2f9b48a5fda0da5bb81c668275c15c7aea380&vid=33a2f9b48a5fda0da5bb81c668275c15c7aea380&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR