Rutaecarpine ≥98.0% (by HPLC, total nitrogen)
Leverantör: TCI EUROPE
Synonyms:
8,13-Dihydroindolo[2',3':3,4]pyrido[2,1-b]quinazolin-5(7H)-one, Rutecarpine, 7,8-Dihydroindolo[2',3':3,4]pyrido[2,1-b]quinazolin-5(13H)-one
R0102-200MG
R0102-1G
TCIAR0102-200MGEA
830
SEK
TCIAR0102-200MG
TCIAR0102-1G
Rutaecarpine ≥98.0% (by HPLC, total nitrogen)
Rutaecarpine
Formel:
C₁₈H₁₃N₃O MW: 287,32 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD00210551 CAS nummer: 84-26-4 Merck Index: 14,08298 |
Specifikation testresultat
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...