13-cis-Retinoic acid ≥97.0% (by HPLC, titration analysis)
Leverantör: TCI
Fara
Synonyms: Isotretinoin, (2Z,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethyl-1-cyclohexen-1-yl)-2,4,6,8-nonatetraenoic Acid, (2Z,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenoic acid
R0088-1G R0088-100MG
TCIAR0088-1GEA 2325 SEK
TCIAR0088-1G TCIAR0088-100MG
13-cis-Retinoic acid ≥97.0% (by HPLC, titration analysis)
13-cis-Retinoic acid
Formel: C₂₀H₂₈O₂
MW: 300.44 g/mol
Smältpunkt: 175°C
Storage Temperature: Frys
MDL Number: MFCD00079542
CAS nummer: 4759-48-2
EINECS: 225-296-0
Merck Index: 14,05228
Order Now
Specifikation testresultat
Appearance Orange to Brown powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %