Ribavirin ≥98.0% (by HPLC, total nitrogen)

Leverantör: TCI EUROPE
Fara

Synonyms: 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1H-1,2,4-triazole-3-carboxamide

R0077-100MG R0077-500MG
TCIAR0077-100MGEA 255 SEK
TCIAR0077-100MG TCIAR0077-500MG
Ribavirin ≥98.0% (by HPLC, total nitrogen)
Ribavirin
Formel: C8H12N4O5
MW: 244,2047 g/mol
MDL Number: MFCD00058564
CAS nummer: 36791-04-5
Merck Index: 14,08198

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Specific rotation [a]20/D -33.0 to -37.0 deg (C=1, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR