Resorufin ≥95.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 7-Hydroxy-3H-phenoxazin-3-one

R0012-1G R0012-5G
TCIAR0012-1GEA 2425 SEK
TCIAR0012-1G TCIAR0012-5G
Resorufin ≥95.0%
Resorufin
Formel: C₁₂H₇NO₃
MW: 213,19 g/mol
MDL Number: MFCD00128991
CAS nummer: 635-78-9
EINECS: 211-241-8

Order Now

Specifikation testresultat

Appearance Dark green to Dark blue to Black powder to crystal
Purity (HPLC) min. 95.0 area%
Lambda max. 569.0 to 573.0 nm (0.01mol/L NaOH sol.)
Absorbance (E1%1cm) min. 2000 (0.01mol/L NaOH sol., 569.0 to 573.0 nm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR