6-Quinoxalinol ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: Quinoxalin-6-ol, 6-Hydroxyquinoxaline

Q0101-200MG Q0101-1G
TCIAQ0101-200MGEA 1475 SEK
TCIAQ0101-200MG TCIAQ0101-1G
6-Quinoxalinol ≥98.0% (by GC, titration analysis)
6-Quinoxalinol
Formel: C₈H₆N₂O
MW: 146,15 g/mol
MDL Number: MFCD07364440
CAS nummer: 7467-91-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=321c4613bb3ea678fd3e1f2bb64fd9c9edacb1fd&vid=321c4613bb3ea678fd3e1f2bb64fd9c9edacb1fd&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Yellow to Brown to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR