Quetiapine hemifumarate ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
ICI-204636, 2-[2-(4-dibenzo[b,f][1,4]thiazepin-11-yl-1-piperazinyl)ethoxy]ethanol bis[(2E)-2-butenedioate], Seroquel
Q0092-5G
Q0092-25G
TCIAQ0092-5GEA
1075
SEK
TCIAQ0092-5G
TCIAQ0092-25G
Quetiapine hemifumarate ≥98.0% (by HPLC, titration analysis)
Quetiapine hemifumarate
Formel:
C₂₁H₂₅N₃O₂S MW: 441,55 g/mol |
MDL Number:
MFCD03423782 CAS nummer: 111974-72-2 Merck Index: 14,08039 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 172.0 to 176.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...