Pifithrin-α hydrobromide ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI
Synonyms: Pifithrin-α HBr, Pifithrin-alpha, 2-(2-Imino-4,5,6,7-tetrahydrobenzothiazol-3-yl)-1-p-tolylethanone Hydrobromide
P2751-10MG P2751-50MG
TCIAP2751-10MGEA 1450 SEK
TCIAP2751-10MG TCIAP2751-50MG
Pifithrin-α hydrobromide ≥98.0% (by HPLC, titration analysis)
Pifithrin-alpha hydrobromide
Formel: C₁₆H₁₈N₂OS·HBr
MW: 286.4·80.9 g/mol
Storage Temperature: Frys
MDL Number: MFCD00417851
CAS nummer: 63208-82-2
Merck Index: 14,07421
Order Now
Specifikation testresultat
Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %