4-Phenyl-2-pyrrolidone ≥98.0%

Leverantör: TCI
Varning

P2727-1G P2727-5G
TCIAP2727-1GEA 510 SEK
TCIAP2727-1G TCIAP2727-5G
4-Phenyl-2-pyrrolidone ≥98.0%
4-Phenyl-2-pyrrolidone
Formel: C₁₀H₁₁NO
MW: 161.21 g/mol
Kokpunkt: 178°C
Storage Temperature: Kylskåp
MDL Number: MFCD01687226
CAS nummer: 1198-97-6

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 74.0 to 78.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR