Pentafluorophenyl Diphenylphosphinate ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
Diphenylphosphinic acid Pentafluorophenyl ester, FDPP, Perfluorophenyl diphenylphosphinate
P2726-1G
P2726-5G
TCIAP2726-1GEA
495
SEK
TCIAP2726-1G
TCIAP2726-5G
Pentafluorophenyl Diphenylphosphinate ≥98.0%
Pentafluorophenyl Diphenylphosphinate
Formel:
C₁₈H₁₀F₅O₂P MW: 384,24 g/mol Kokpunkt: 153°C |
MDL Number:
MFCD00192380 CAS nummer: 138687-69-1 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 55.0 to 59.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...