PEG3-carboxylic acid tert-Butyl ester ≥97.0%
Leverantör: TCI
Synonyms: tert-Butyl 3-(2-(2-hydroxyethoxy)ethoxy)propanoate, 3-[2-(2-Hydroxyethoxy)ethoxy]propanoic acid tert-Butyl ester, Hydroxy-PEG2-(CH2)2-Boc, tert-Butyl 3-[2-(2-Hydroxyethoxy)ethoxy]propanoate, 9-Hydroxy-4,7-dioxanonanoic acid tert-Butyl ester, tert-Butyl 9-Hydroxy-4,7-dioxanonanoate
P2719-1G P2719-5G
TCIAP2719-1GEA 1075 SEK
TCIAP2719-1G TCIAP2719-5G
PEG3-carboxylic acid tert-Butyl ester ≥97.0%
PEG3-carboxylic acid tert-Butyl ester
Formel: C₁₁H₂₂O₅
MW: 234.29 g/mol
Storage Temperature: Frys
MDL Number: MFCD20926380
CAS nummer: 133803-81-3
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %