Polyethylene glycol diacrylate (PEGDA) n=∼9 stabilised

Leverantör: TCI EUROPE
Fara

Synonyms: PEG hydrogel, Polyethylene glycol diacrylate, Poly(ethylene glycol) diacrylate, PEG diacrylate, PEGDA, PEG bisacrylate

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

P2708-25G P2708-500G
TCIAP2708-25GEA 285 SEK
TCIAP2708-25G TCIAP2708-500G
Polyethylene glycol diacrylate (PEGDA) n=∼9 stabilised
Polyethylene glycol diacrylate (PEGDA)
Formel: CH₂=COC(OCH₂CH₂)nOCOC=CH₂
Flampunkt: 104°C
MDL Number: MFCD00081876
CAS nummer: 26570-48-9

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
The average value of n 8.5 to 9.5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR