Phenyl-p-benzoquinone ≥96.0%

Leverantör: TCI EUROPE

Synonyms: 2-Phenyl-1,4-benzoquinone, [1,1'-Biphenyl]-2,5-dione

P2674-1G P2674-5G
TCIAP2674-1GEA 335 SEK
TCIAP2674-1G TCIAP2674-5G
Phenyl-p-benzoquinone ≥96.0%
Phenyl-p-benzoquinone
Formel: C₁₂H₈O₂
MW: 184,194 g/mol
MDL Number: MFCD00001599
CAS nummer: 363-03-1
EINECS: 206-654-5

Order Now

Specifikation testresultat

Appearance White to Brown to Dark purple powder to crystal
Purity (GC) min. 96.0 %
Melting point 111.0 to 115.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR