5-Phenyl-1H-[1,2,4]triazol-3-ylamine ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 5-Phenyl-1H-1,2,4-triazol-3-amine, 3-Phenyl-1H-1,2,4-triazol-5-amine

P2658-1G P2658-5G
TCIAP2658-1GEA 1725 SEK
TCIAP2658-1G TCIAP2658-5G
5-Phenyl-1H-[1,2,4]triazol-3-ylamine ≥98.0% (by GC, titration analysis)
5-Phenyl-1H-[1,2,4]triazol-3-ylamine
Formel: C₈H₈N₄
MW: 160,18 g/mol
MDL Number: MFCD00086332
CAS nummer: 4922-98-9

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 188.0 to 192.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR