2-Phenylimidazo[1,2-a]pyridine ≥98.0%
Leverantör: TCI EUROPE
P2632-1G
P2632-5G
TCIAP2632-1GEA
1300
SEK
TCIAP2632-1G
TCIAP2632-5G
2-Phenylimidazo[1,2-a]pyridine ≥98.0%
2-Phenylimidazo[1,2-a]pyridine
Formel:
C₁₃H₁₀N₂ MW: 194,24 g/mol Kokpunkt: 245°C |
MDL Number:
MFCD00219254 CAS nummer: 4105-21-9 |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 133.0 to 137.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...