Protopine ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 7-Methyl-6,7,8,9-tetrahydro-[1,3]dioxolo[4',5':5,6]benzo[1,2-c][1,3]dioxolo[5',4':4,5]benzo[1,2-g]azecin-15(16H)-one

P2611-5MG P2611-25MG P2611-100MG
TCIAP2611-5MGEA 925 SEK
TCIAP2611-5MG TCIAP2611-25MG TCIAP2611-100MG
Protopine ≥98.0%
Protopine
Formel: C₂₀H₁₉NO₅
MW: 353,37 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00022322
CAS nummer: 130-86-9
EINECS: 204-999-6
UN: 1544
ADR: 6.1,III
Merck Index: 14,07896

Order Now

Specifikation testresultat

Appearance White to Light gray to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 201.0 to 205.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR