Propoxybenzene ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Phenyl propyl ether
P2607-25ML
P2607-5ML
TCIAP2607-25MLEA
3875
SEK
TCIAP2607-25ML
TCIAP2607-5ML
Propoxybenzene ≥98.0%
Propoxybenzene
Formel:
C₉H₁₂O MW: 136,19 g/mol Kokpunkt: 190°C Smältpunkt: -28°C |
MDL Number:
MFCD00039935 CAS nummer: 622-85-5 EINECS: 210-756-5 |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...