5'-Phenyl-[1,1':3',1''-terphenyl]-4-amine ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 5'-Phenyl-m-terphenyl-4-amine

P2589-1G P2589-5G
TCIAP2589-1GEA 1275 SEK
TCIAP2589-1G TCIAP2589-5G
5'-Phenyl-[1,1':3',1''-terphenyl]-4-amine ≥98.0%
5'-Phenyl-[1,1':3',1''-terphenyl]-4-amine
Formel: C₂₄H₁₉N
MW: 321,42 g/mol
MDL Number: MFCD22581309
CAS nummer: 343239-58-7

Order Now

Specifikation testresultat

Appearance White to Gray to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 152.0 to 156.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR