Pentafluorobenzyl methacrylate ≥98.0% stabilised

Leverantör: TCI
Varning

Synonyms: Methacrylic acid Pentafluorobenzyl ester

Stabilization: Stabilised med BHT/Ionol

P2546-5G P2546-25G
TCIAP2546-5GEA 1750 SEK
TCIAP2546-5G TCIAP2546-25G
Pentafluorobenzyl methacrylate ≥98.0% stabilised
Pentafluorobenzyl methacrylate
Formel: C₁₁H₇F₅O₂
MW: 266.17 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00042329
CAS nummer: 114859-23-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2d36f51bc5d87082a8edfecc836457281d6061f1&vid=2d36f51bc5d87082a8edfecc836457281d6061f1&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White or Colorless to Yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR