Potassium tricyanomethanide ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
P2538-1G
P2538-5G
TCIAP2538-1GEA
1150
SEK
TCIAP2538-1G
TCIAP2538-5G
Potassium tricyanomethanide ≥98.0%
Potassium tricyanomethanide
Formel:
C₄KN₃ Smältpunkt: 298°C |
MDL Number:
MFCD00058850 CAS nummer: 34171-69-2 UN: 3439 ADR: 6.1,II |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Ion exchange titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...