Poly(3-hexylthiophene-2,5-diyl) regioregular
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
P3HT
P2513-100MG
P2513-500MG
TCIAP2513-100MGEA
2650
SEK
TCIAP2513-100MG
TCIAP2513-500MG
Poly(3-hexylthiophene-2,5-diyl) regioregular
Poly(3-hexylthiophene-2,5-diyl)
MW:
196,352 g/mol |
MDL Number:
MFCD26958730 CAS nummer: 125321-66-6 |
Specifikation testresultat
Hole Mobility (mu FET) | min 0.10cm2/Vs (OTS Si/SiO2 substrate) |
Palladium | max. 100 ppm |
Molecular weight of number average | 27,000 to 45,000 |
Stereoregularity | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...