Potassium bis(fluorosulphonyl)imide ≥95.0%
Leverantör: TCI
|
Fara
|
Synonyms:
Potassium bis(fluorosulphonyl)amide
P2509-5G
P2509-25G
TCIAP2509-5GEA
565
SEK
TCIAP2509-5G
TCIAP2509-25G
Potassium bis(fluorosulphonyl)imide ≥95.0%
Potassium bis(fluorosulphonyl)imide
Formel:
F₂KNO₄S₂ MW: 219.23 g/mol |
MDL Number:
MFCD17018950 CAS nummer: 14984-76-0 UN: 1759 ADR: 8 II |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Ion exchange titration) | min. 95.0 % |
Melting point | 99.0 to 103.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...