4-(1H-Pyrazol-1-yl)benzoic acid ≥98.0%

Leverantör: TCI EUROPE
Varning

P2499-1G P2499-5G
TCIAP2499-1GEA 615 SEK
TCIAP2499-1G TCIAP2499-5G
4-(1H-Pyrazol-1-yl)benzoic acid ≥98.0%
4-(1H-Pyrazol-1-yl)benzoic acid
Formel: C₁₀H₈N₂O₂
MW: 188,18 g/mol
MDL Number: MFCD02682056
CAS nummer: 16209-00-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e4613c5a472a8a9de5dc884aca1b95e1ecd08da&vid=0e4613c5a472a8a9de5dc884aca1b95e1ecd08da&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR