Piperazine dihydrochloride ≥98.0% (by total nitrogen and titration analysis)
Leverantör: TCI
Fara
Synonyms: Dihydrochloride salt of diethylenediamine, Piperazinium chloride, Piperazinium dichloride
P2491-1G P2491-5G
TCIAP2491-1GEA 630 SEK
TCIAP2491-1G TCIAP2491-5G
Piperazine dihydrochloride ≥98.0% (by total nitrogen and titration analysis)
Piperazine dihydrochloride
Formel: C₄H₁₀N₂·2HCl
MW: 159.05 g/mol
Smältpunkt: 320°C
MDL Number: MFCD00012754
CAS nummer: 142-64-3
EINECS: 205-551-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70985ed3f21e8a01127b1ce847ab613adfa90990&vid=70985ed3f21e8a01127b1ce847ab613adfa90990&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %