1H-Pyrazole-4-boronic acid (contains varying amounts of Anhydride)

Leverantör: TCI EUROPE
Varning

P2478-1G P2478-5G
TCIAP2478-1GEA 550 SEK
TCIAP2478-1G TCIAP2478-5G
1H-Pyrazole-4-boronic acid (contains varying amounts of Anhydride)
1H-Pyrazole-4-boronic acid
Formel: C₃H₅BN₂O₂
MW: 111,9000 g/mol
MDL Number: MFCD07368251
CAS nummer: 763120-58-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc8da9ee82f4cb57c3673dd0814dc01651b54ca1&vid=bc8da9ee82f4cb57c3673dd0814dc01651b54ca1&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (Neutralization titration) 98.0 to 119.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR