10-Phenylphenothiazine ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
10-Phenyl-10H-phenothiazine
P2470-1G
P2470-5G
TCIAP2470-1GEA
1700
SEK
TCIAP2470-1G
TCIAP2470-5G
10-Phenylphenothiazine ≥98.0%
10-Phenylphenothiazine
Formel:
C₁₈H₁₃NS MW: 275,37 g/mol Kokpunkt: 223°C |
MDL Number:
MFCD00156238 CAS nummer: 7152-42-3 |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 93.0 to 97.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...